• USA Home
  • 34802 - HYDRANAL®-Water-in-methanol 5.0

EMAIL THIS PAGE TO A FRIEND
34802 Fluka

HYDRANAL®-Water-in-methanol 5.0

for volumetric back titration according to Karl Fischer, 5.00 mg/mL±0.05 mg/mL in methanol

Synonym: Methanol solution

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Karl Fischer Titration Reagents, One-component reagents, Titration,
quality   for volumetric back titration according to Karl Fischer
concentration   5.00 mg/mL±0.05 mg/mL in methanol
suitability   use tested

Description

General description

Visit our HYDRANAL® center to learn more.

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

HYDRANAL is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1230 3/PG 2
WGK Germany 
1
RTECS 
PC1400000
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?