Popular Documents: Datasheet (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, KF Reagents for Special Applications, Karl Fischer Titration Reagents (HYDRANAL), One-component reagents, |
| suitability | use tested |
| compatibility | reagent for volumetric one-component KF titration in aldehydes and ketones (methanol-free titrating agent) |
To be used with: see related products
Visit our HYDRANAL center to learn more.
4×2.5 L in aluminum bottle
6×500 mL in ampule
6×1 L in ampule
Water equivalent 4.5-5.5 mg H2O/mL
HYDRANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Analysis of Bioethanol - Detection of Chloride and Sulfate with Dication and Trication Solutions
Customers Also Viewed
reagent for volumetric one-component KF titration (methanol-free titrating agent)
reagent for volumetric one-component KF titration (methanol-free titrating agent)
reagent for volumetric one-component KF titration (methanol-free titrating agent)
reagent for volumetric one-component KF titration in aldehydes and ketones (working medium)
reagent for volumetric one-component KF titration in aldehydes and ketones (working medium)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?