34821 Fluka

HYDRANAL®-Coulomat CG-K

reagent for coulometric KF titration in ketones (catholyte solution)

DOWNLOAD MSDS (PDF)

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Coulometric Reagents, KF Reagents for Special Applications, Karl Fischer Titration Reagents (HYDRANAL),
composition   methylformamide
suitability   use tested
compatibility   reagent for coulometric KF titration in ketones (catholyte solution)

Description

Application

To be used with: see related products

General description

Visit our HYDRANAL center to learn more.

Packaging

10 × 5 mL ampules

Legal Information

HYDRANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Protocols & Applications

Analysis of Bioethanol - Detection of Chloride and Sulfate with Dication and Trication Solutions

Price and Availability

Customers Also Viewed

Dibutyl phosphite

technical, ≥95% (GC)

reagent for coulometric KF titration (anolyte solution), for cells with diaphragm

reagent for coulometric KF titration (anolyte solution), for cells with and without diaphragm

reagent for coulometric KF titration in ketones (anolyte solution), for cells with and without diaphragm

reagent for coulometric KF titration (catholyte solution)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?