• USA Home
  • 34849 - HYDRANAL®-Water Standard 10.0

EMAIL THIS PAGE TO A FRIEND
34849 Fluka

HYDRANAL®-Water Standard 10.0

standard for Karl Fischer titration (water content 10 mg/g = 1.0%), exact value on report of analysis

Purchase

Properties

Related Categories Analytical Reagents, Analytical Standards, Analytical/Chromatography, Applications, Biodiesel,
composition   1-butanol
density   1.00 g/mL at 20 °C
suitability   suitable for
  use tested
compatibility   standard for Karl Fischer titration (water content 10 mg/g = 1.0%), exact value on report of analysis

Description

General description

This standard is tested against NIST SRM 2890

Visit our HYDRANAL® center to learn more.

Packaging

Box contains 10 glass ampules of 8 mL.

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

HYDRANAL is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
2
Flash Point(F) 
86 °F
Flash Point(C) 
30 °C
Protocols & Articles

Articles

Titer Determination and System Check for Karl Fischer Titration

Calibration, validation and control of analytical instruments and reagents are central requirements in most quality management systems. In the case of Karl Fischer (KF) titration, regular titer deter...
Andrea Felgner
Analytix 2013, Volume 5
Keywords: Karl Fischer titrations, Titrations

Related Content

Water Standards Quality Control [VIDEO]

This Karl Fischer technical tip video provides factors to consider when selecting an appropriate water standard for your application. The options of pure water, sodium tartrate dihydrate, HYDRANAL® w...
Keywords: Karl Fischer titrations, Titrations

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules
34425 Water Standard 10.0 CRM, certified reference material (for Karl Fischer titration, water content 10 mg/g = 1.0%, exact value on CoA)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?