34966 Fluka

Methanol

LC-MS CHROMASOLV®

DOWNLOAD MSDS (PDF)

Synonym: Methyl alcohol

Properties

Related Categories 3-MCPD, Acrylamide, Adulterants, Allergens, Amber Glass Bottles,
grade   LC-MS CHROMASOLV®
vapor density   1.11 (vs air)
vapor pressure   410 mmHg ( 50 °C)
  97.68 mmHg ( 20 °C)
assay   ≥99.9%
autoignition temp.   725 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   36 %
impurities   ≤0.0005% free alkali (as NH3)
  ≤0.0005% non-volatile matter
  ≤0.001% free acid (as HCOOH)
  ≤0.02% water (Karl Fischer)
  ≤1 ppb fluorescence (quinine) at 254 nm
  ≤1 ppb fluorescence (quinine) at 365 nm
transmittance   210 nm, ≥30%
  220 nm, ≥50%
  230 nm, ≥75%
  260 nm, 98%
refractive index   n20/D 1.329(lit.)
bp   64.7 °C(lit.)
mp   −98 °C(lit.)
density   0.791 g/mL at 25 °C(lit.)
cation traces   Ag: ≤0.1 mg/kg
  Al: ≤0.5 mg/kg
  Ba: ≤0.1 mg/kg
  Ca: ≤0.1 mg/kg
  Cd: ≤0.05 mg/kg
  Co: ≤0.02 mg/kg
  Cr: ≤0.02 mg/kg
  Cu: ≤0.01 mg/kg
  Fe: ≤0.1 mg/kg
  K: ≤0.1 mg/kg
  Mg: ≤0.1 mg/kg
  Mn: ≤0.01 mg/kg
  Na: ≤0.1 mg/kg
  Ni: ≤0.02 mg/kg
  Pb: ≤0.02 mg/kg
  Sn: ≤0.1 mg/kg
  Zn: ≤0.1 mg/kg
abs.   HPLC-gradient/254 nm ≤5 mAU
suitability   passes test for suitability for LC-MS

Description

Application

Watch a 6-minute presentation about the advantages of our High Purity Solvents for Sensitive LC-MS Analysis

Packaging

1, 6×1, 2.5, 4×2.5, 4×4 L in glass bottle

Preparation Note

filtered through a 0.2 μm filter

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Customers Also Viewed

2-Propanol

LC-MS CHROMASOLV®

Acetonitrile

LC-MS CHROMASOLV®

Methanol

CHROMASOLV®, for HPLC, ≥99.9%

Methanol

LC-MS Ultra CHROMASOLV®, tested for UHPLC-MS

Methanol

CHROMASOLV®, gradient grade, for HPLC, ≥99.9%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?