34972 Fluka

Ethyl acetate

LC-MS CHROMASOLV®

DOWNLOAD MSDS (PDF)

Synonym: EtOAc

  • CAS Number 141-78-6

  • Linear Formula CH3COOC2H5

  • Molecular Weight 88.11

  •  Beilstein Registry Number 506104

  •  EC Number 205-500-4

  •  MDL number MFCD00009171

  •  PubChem Substance ID 24861703

Properties

Related Categories Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, CHROMASOLV LC-MS, Chromatography Reagents & Solvents,
grade   LC-MS CHROMASOLV®
vapor density   3 (20 °C, vs air)
vapor pressure   73 mmHg ( 20 °C)
assay   ≥99.7% (GC)
autoignition temp.   801 °F
expl. lim.   2.2-11.5 %, 38 °F
impurities   ≤0.0005% free alkali (as NH3)
  ≤0.0005% non-volatile matter
  ≤0.03% water (Karl Fischer)
transmittance   260 nm, ≤50%
  300 nm, ≤98%
refractive index   n20/D 1.3720(lit.)
bp   76.5-77.5 °C(lit.)
mp   −84 °C(lit.)
density   0.902 g/mL at 25 °C(lit.)
cation traces   Ca: ≤0.1 mg/kg
  K: ≤0.1 mg/kg
  Mg: ≤0.1 mg/kg
  Na: ≤0.1 mg/kg
suitability   passes test for suitability for LC-MS

Description

Packaging

1, 2.5 L in glass bottle

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Customers Also Viewed

Ethyl acetate

anhydrous, 99.8%

Ethyl acetate

CHROMASOLV® Plus, for HPLC, 99.9%

Ethyl acetate

CHROMASOLV®, for HPLC, ≥99.8%

Ethyl acetate

CHROMASOLV®, for HPLC, ≥99.7%

2000 μg/mL in methanol, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?