Synonym:
4-Amino-N-
CAS Number 68-35-9
Empirical Formula (Hill Notation) C10H10N4O2S
Molecular Weight 250.28
Beilstein Registry Number 6733588
EC Number 200-685-8
MDL number MFCD00006065
PubChem Substance ID 24861734
| Related Categories | 1694 Pharmaceuticals & Personal Care Products, APIs (Active Pharmaceutical Ingredients) & Metabolites, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, |
| grade | VETRANAL™, analytical standard |
| mp | 253 °C (dec.)(lit.) |
| suitability | suitable for 1694 per US EPA |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?