CAS Number 7778-50-9
Linear Formula K2Cr2O7
Molecular Weight 294.18
MDL number MFCD00011367
PubChem Substance ID 24861758
| Related Categories | Analytical Reagents, Analytical/Chromatography, By Reference Material, Chemical Synthesis, Chromates, |
| quality | volumetric |
| concentration |
|
| factor | 1.000 ±0.001 |
Visit our Titration Center to learn more.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?