CAS Number 7722-64-7
Empirical Formula (Hill Notation) KMnO4
Molecular Weight 158.03
MDL number MFCD00011364
PubChem Substance ID 24861763
| Related Categories | Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Most common compounds for volumetric titration, N - R, |
| quality | volumetric |
| concentration | 0.2 M KMnO4 (1N) |
Visit our Titration Center to learn more.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?