Synonym:
(2,4′-Dichlorodiphenyl)dichloroethane,
1-
CAS Number 53-19-0
Linear Formula (ClC6H4)2CHCHCl2
Molecular Weight 320.04
Beilstein Registry Number 2056007
EC Number 200-166-6
MDL number MFCD00000850
PubChem Substance ID 24861830
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Bioactive Small Molecules, |
| grade | PESTANAL®, analytical standard |
| Gene Information | rat ... Ar(24208) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?