Synonym: 2,4,4′-PCB, 2,4,4′-Trichlorobiphenyl
CAS Number 7012-37-5
Empirical Formula (Hill Notation) C12H7Cl3
Molecular Weight 257.54
Ballschmiter Number 28
Beilstein Registry Number 1956846
EC Number 230-293-2
MDL number MFCD00055554
PubChem Substance ID 24861856
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Aroclors, PCBs, and Dioxins, Chromatography, |
| grade | analytical standard, for environmental analysis |
Customers Also Viewed
1000 μg/mL in isooctane (each solution), analytical standard
analytical standard, for environmental analysis, 6 PCB according to TrinkwV (BRD) method, in isooctane
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?