Synonym: 2,2′,4,4′,5,5′-Hexachlorobiphenyl, 2,2′,4,4′,5,5′-PCB
CAS Number 35065-27-1
Empirical Formula (Hill Notation) C12H4Cl6
Molecular Weight 360.88
Ballschmiter Number 153
Beilstein Registry Number 1990497
MDL number MFCD00055535
| Related Categories | 2000/60/EC, 8000 Series Solidwaste Methods, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard, for environmental analysis |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?