| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, ICP-OES/-MS, ICP/DCP Standard Solutions, |
| concentration | 10,000 μg/mL Mg in 2% HNO3 (Exact Mg concentration on label) |
| density | 1.05 g/mL at 25 °C |
Customers Also Viewed
TraceCERT®, 1000 mg/L Mg in nitric acid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?