CAS Number 218-01-9
Empirical Formula (Hill Notation) C18H12
Molecular Weight 228.29
Beilstein Registry Number 1909297
EC Number 205-923-4
MDL number MFCD00003698
PubChem Substance ID 24861925
| Related Categories | All PAH Standards, Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard, for environmental analysis |
| bp | 448 °C(lit.) |
| mp | 252-254 °C(lit.) |
Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?