• USA Home
  • 35990 - 2,3-Dichloro-1,4-naphthoquinone

EMAIL THIS PAGE TO A FRIEND

35990 Fluka

2,3-Dichloro-1,4-naphthoquinone

for spectrophotometric det. of hydrazides, ≥98.0%

Synonym: Dichlon

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Spectroscopy, UV/Vis Reagents, UV/Visible (UV/VIS) Spectroscopy More...
description   algaecide
assay   ≥98.0% (AT)
  ≥98.0%
quality   for spectrophotometric det. of hydrazides
mp   192-195 °C
  194-197 °C(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
26-60-61
RIDADR 
UN 2811 6.1/PG 3
WGK Germany 
3
RTECS 
QL7525000

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Irreversible inhibition of rat hepatic glutathione S-transferase isoenzymes by a series of structurally related quinones. Vos RM, Van Ommen B, Hoekstein MS, et al. Chem. Biol. Interact. 71(4), 381-92, (1989)

Loading...


[Interaction of methimazole and quinones. Spectrophotometric determination of methimazole in blood]. Gallo N, Bianco P, Tapino R, et al. Minerva Med. 74(16), 875-7, (1983)

Loading...


Indoleamine 2,3-dioxygenase is the anticancer target for a novel series of potent naphthoquinone-based inhibitors. Sanjeev Kumar et al J. Med. Chem. 51, 1706-18, (2008)

Loading...


Toxicology in the fast lane: application of high-throughput bioassays to detect modulation of key enzymes and receptors. Morisseau C, Merzlikin O, Lin A, et al. Environ. Health Perspect. 117(12), 1867-72, (2009)

Loading...


2,3-Disubstituted-1,4-naphthoquinones, 12H-benzo[b]phenothiazine-6,11-diones and related compounds: synthesis and biological evaluation as potential antiproliferative and antifungal agents. Vishnu K Tandon et al Eur. J. Med. Chem. 44, 1086-92, (2009)

Loading...


A redox cycling mechanism of action for 2,3-dichloro-1,4-naphthoquinone with mitochondrial membranes and the role of sulfhydryl groups. Pritsos CA and Pardini RS Biochem. Pharmacol. 33(23), 3771-7, (1984)

Loading...


Metabolic consequences of dietary 2,3-dichloro-1,4-naphthoquinone (CNQ) in the rat. Alteration in anti-oxidant enzyme activities. Pritsos CA, Aaronson LM, and Pardini RS Biochem. Pharmacol. 35(7), 1131-5, (1986)

Loading...


A role for glutathione disulfide as a scavenger of oxygen radicals produced by 2,3-dichloro-1,4-naphthoquinone. Pritsos CA and Pardini RS Res. Commun. Chem. Pathol. Pharmacol. 42(2), 271-80, (1983)

Loading...


The 1,4-naphthoquinone scaffold in the design of cysteine protease inhibitors. Cláudia Valente et al Bioorg. Med. Chem. 15, 5340-50, (2007)

Loading...


Involvement of superoxide in the interaction of 2,3-dichloro-1,4-naphthoquinone with mitochondrial membranes. Pritsos CA, Jensen DE, Pisani D, et al. Arch. Biochem. Biophys. 217(1), 98-109, (1982)

Loading...


Development and validation of spectrophotometric methods for determination of fluoxetine, sertraline, and paroxetine in pharmaceutical dosage forms. Darwish IA J. AOAC Int. 88(1), 38-45, (2005)

Loading...


Incorporation of a high potential quinone reveals that electron transfer in Photosystem I becomes highly asymmetric at low temperature. Mula S, Savitsky A, Möbius K, et al. Photochem. Photobiol. Sci. 11(6), 946-56, (2012)

Loading...


Interaction of 2,3-dichloro-1,4-naphthoquinone with n-butylamine in halocarbon solvents. Neelgund GM and Budni ML Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 61(8), 1729-35, (2005)

Loading...


New quinone-amino acid conjugates linked via a vinylic spacer. Alnabari M and Bittner S Amino Acids 20(4), 381-7, (2001)

Loading...


Modulation of hepatic cytochrome P-450 and DT-diaphorase by oral and sub-cutaneous administration of the pro-oxidant fungicide dichlone (2,3-dichloro-1,4-naphthoquinone). Elliott AJ and Pardini RS Bull. Environ. Contam. Toxicol. 41(2), 164-71, (1988)

Loading...


Spectroscopic and theoretical studies on the nucleophilic substitution of 2,3-dichloronaphthoquinone with para-substituted anilines in solid state via initial charge transfer complexation. Satheshkumar A and Elango KP Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 98, 378-83, (2012)

Loading...


Study of a reaction between 2,3-dichloro-1,4-naphthoquinone and N,N'-diphenyl thiourea involving an EDA adduct as intermediate. Datta K and Mukherjee AK Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 60(7), 1641-7, (2004)

Loading...


Charge transfer interaction of 4-acetamidophenol (paracetamol) with 2,3-dichloro-1,4-naphthoquinone: a study in aqueous ethanol medium by UV-vis spectroscopic and DFT methods. Saha A, Tiwary AS, and Mukherjee AK Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 71(3), 835-40, (2008)

Loading...


Dichlone-induced oxidative stress in a model insect species, Spodoptera eridania. Ahmad S, Zaman K, MacGill RS, et al. Arch. Environ. Contam. Toxicol. 29(4), 442-8, (1995)

Loading...


Inhibition of liver glycolysis in rats by dietary dichlone (2,3-dichloro-1,4-naphthoquinone). Pritsos CA, Pisani DE, and Pardini RS Bull. Environ. Contam. Toxicol. 35(1), 23-8, (1985)

Loading...


Bacterial growth response to photoactive quinones. Vaughan PP, Novotny P, Haubrich N, et al. Photochem. Photobiol. 86(6), 1327-33, (2010)

Loading...


[Reactions between dialkylamine drugs, 2,3-dichloro-1,4-naphthoquinone and acetaldehyde]. Kallmayer HJ and Thierfelder B Pharmazie 57(8), 530-4, (2002)

Loading...


[Reactions of isopropyl-alkylamine with 2,3-dichloro-1,4-naphthoquinone and 2,3-dichloro-1,4-naphthoquinone/acetaldehyde]. Kallmayer HJ and Thierfelder B Pharmazie 58(2), 104-7, (2003)

Loading...


[Reactions of trialkylamine compounds with 2,3-dichloro-1,4-naphthoquinone]. Kallmayer HJ and Thierfelder B Pharmazie 57(9), 648-9, (2002)

Loading...


[Preparation of water-soluble hydronaphthoquinone derivatives and its application to the determination of hydrogen peroxide in clinical chemistry]. Ohta S, Hatakeyama S, Aono C, et al. Yakugaku Zasshi 112(8), 585-91, (1992)

Loading...


Merck 14,3045

Beil. 7,IV,2426

Corp MSDS 1 (1), 1146:A / FT-IR 1 (2), 80:D / FT-IR 2 (2), 2485:B / IR-Spectra (2), 785:A / IR-Spectra (3), 897:E / NMR-Reference 2 (2), 86:D / RegBook 1 (2), 1683:K / Sax 6, 953 / Structure Index 1, 267:D:1

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?