Synonym:
Bis(5-
CAS Number 97-23-4
Linear Formula CH2[C6H3(Cl)OH]2
Molecular Weight 269.12
Beilstein Registry Number 1884514
EC Number 202-567-1
MDL number MFCD00002322
PubChem Substance ID 24862123
| Related Categories | Algicides, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
| mp | 168-172 °C(lit.) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?