Synonym: Alkaline potassium sodium tartrate, Potassium sodium tartrate solution, alkaline
MDL number MFCD00012467
| grade | Reag. Ph. Eur. |
| for determination of sugar | |
| density | 1.23-1.25 g/mL at 20 °C |
| suitability | in accordance for application |
Customers Also Viewed
for the qualitative determination of sugars
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?