Synonym: Alphamethrine
CAS Number 52315-07-8
Empirical Formula (Hill Notation) C22H19Cl2NO3
Molecular Weight 416.30
Beilstein Registry Number 8444118
EC Number 257-842-9
MDL number MFCD00055328
| Related Categories | Acaricides, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
| description | mixture of isomers |
| refractive index |
n |
| suitability | passes test for identity (NMR) |
Commission Regulation (EU) No 37/2010 of 22 December 2009 on pharmacologically active substances and their classification regarding maximum residue limits in foodstuffs of animal origin
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?