CAS Number 121-75-5
Empirical Formula (Hill Notation) C10H19O6PS2
Molecular Weight 330.36
Beilstein Registry Number 1804525
EC Number 204-497-7
MDL number MFCD00036233
PubChem Substance ID 24862222
| Related Categories | 2000/60/EC, Acaricides, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, |
| grade | PESTANAL®, analytical standard |
| storage temp. | 2-8°C |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?