Synonym:
2-Chloro-N-
CAS Number 51218-45-2
Empirical Formula (Hill Notation) C15H22ClNO2
Molecular Weight 283.79
Beilstein Registry Number 8396147
EC Number 257-060-8
MDL number MFCD00055293
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Amide, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
| refractive index |
n |
| suitability | passes test for identity (NMR) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?