Synonym: O,O-Dimethyl O-(4-nitrophenyl) phosphorothioate, Methyl parathion
CAS Number 298-00-0
Empirical Formula (Hill Notation) C8H10NO5PS
Molecular Weight 263.21
Beilstein Registry Number 8905814
EC Number 206-050-1
MDL number MFCD00041813
PubChem Substance ID 24862271
| Related Categories | 2000/60/EC, Acaricides, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, |
| grade | PESTANAL®, analytical standard |
| storage temp. | 2-8°C |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
1000 μg/mL in acetone, analytical standard
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?