CAS Number 108-70-3
Empirical Formula (Hill Notation) C6H3Cl3
Molecular Weight 181.45
Beilstein Registry Number 1635233
EC Number 203-608-6
MDL number MFCD00000585
PubChem Substance ID 24862537
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
| bp | 208 °C(lit.) |
| mp | 56-60 °C(lit.) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?