Synonym: N-Benzyl-N-isopropylpivalamide
CAS Number 35256-85-0
Empirical Formula (Hill Notation) C15H23NO
Molecular Weight 233.35
Beilstein Registry Number 2838127
EC Number 252-470-3
MDL number MFCD00145351
PubChem Substance ID 24862554
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Amide, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?