Synonym:
1,1,1-
CAS Number 50-29-3
Beilstein Registry Number 1882657
EC Number 200-659-6
MDL number MFCD00000802
PubChem Substance ID 24862678
| Related Categories | 2000/60/EC, Acaricides, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, |
| grade | PESTANAL®, analytical standard |
| concentration | 100 ng/μL in methanol |
| storage temp. | 2-8°C |
| Gene Information |
mouse ... Esr1(13982) rat ... Ar(24208) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?