Synonym: Indicator sticks, special
| Related Categories | Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Test Papers/Sticks, pH Papers/Sticks More... |
| quality | non-bleeding |
| coloring against scale | complies |
| pH | 0.0-6.0 |
box with 100 strips
Customers Also Viewed
non-bleeding
pH 0.5-5.5
non-bleeding, pH 4.5-10.0
non-bleeding, pH 7.0-14.0
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?