37203 Fluka
Ascorbic acid test sticks Quantofix®
50-2000 mg/L, (vitamin C)
DOWNLOAD MSDS (PDF)
Description
General description
Contents:
Tube with 100 test sticks.
Storage:
Avoid exposing the strips to sunlight and moisture. Keep container cool and dry (store below +30 °C).
Gradation:
0 - 50 - 100 - 200 - 300 - 500 - 1000 - 2000 mg/L Ascorbic acid / Vitamin C
Legal Information
Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG
Price and Availability
Customers Also Viewed
<SC>L</SC>-Ascorbic acid
BioXtra, ≥99.0%, crystalline
Quantofix<SUP>®</SUP> test strips
Ascorbic acid (Vit.C)
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?