• USA Home
  • 37761 - 2,3-Dihydroxynaphthalene

EMAIL THIS PAGE TO A FRIEND

37761 Fluka

2,3-Dihydroxynaphthalene

technical, ≥90% (HPLC)

Synonym: 2,3-Naphthalenediol

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
grade   technical
assay   ≥90% (HPLC)
mp   161-165 °C(lit.)
  161-165 °C
Gene Information   human ... BAD(572)

Description

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

1,2-<WBR>Dihydroxynaphthalene

technical grade

1,3-<WBR>Dihydroxynaphthalene

≥99%, crystalline

2,3-<WBR>Dihydroxynaphthalene

≥98.0% (HPLC), purified by sublimation

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
Safety Statements (Europe) 
26
WGK Germany 
3
RTECS 
QJ4750000

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Selective antiproliferative activity of hydroxynaphthyl-beta-D-xylosides. Mårten Jacobsson et al J. Med. Chem. 49, 1932-8, (2006)

Loading...


Reactions of linoleic acid peroxyl radicals with phenolic antioxidants: a pulse radiolysis study. Erben-Russ M, Bors W, and Saran M Int. J. Radiat. Biol. Relat. Stud. Phys. Chem. Med. 52(3), 393-412, (1987)

Loading...


Hydrolytic metal with a hydrophobic periphery: titanium(IV) complexes of naphthalene-2,3-diolate and interactions with serum albumin. Tinoco AD, Eames EV, Incarvito CD, et al. Inorg. Chem. 47(18), 8380-90, (2008)

Loading...


A novel "pro-sensitizer" based sensing of enzymes using Tb(III) luminescence in a hydrogel matrix. Bhowmik S and Maitra U Chem. Commun. (Camb.) 48(38), 4624-6, (2012)

Loading...


The multidrug efflux regulator TtgV recognizes a wide range of structurally different effectors in solution and complexed with target DNA: evidence from isothermal titration calorimetry. Guazzaroni ME, Krell T, Felipe A, et al. J. Biol. Chem. 280(21), 20887-93, (2005)

Loading...


Regulation of rat liver phenylalanine hydroxylase. Kinetic properties of the enzyme's iron and enzyme reduction site. Shiman, R., et al. J. Biol. Chem. 269, 24637, (1994)

Loading...


The syntheses and characterizations of molybdenum(VI) complexes with catechol and 2,3-dihydroxynaphthalene, and the structure-effect relationship in their in vitro anticancer activities. Feng J, Lu XM, Wang G, et al. Dalton Trans. 41(28), 8697-702, (2012)

Loading...


Redox cycling by cytochrome P-450. Besse P, Modi S, Boyle JM, et al. Biochem. Soc. Trans. 25(4), S579, (1997)

Loading...


Terminal group effects on the fluorescence spectra of europium(III) nitrate complexes with a family of amide-based 2,3-dihydroxynaphthalene derivatives. Lei KW and Liu WS Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 65(1), 187-90, (2006)

Loading...


[The synthesis, 1H NMR and IR study of [(n-Bu)4N]2[Mo2O5(OC10H6O)2] and [(n-Bu)4N]2[Mo4O10 (OC10H6O)2(OCH3)2]]. Lu XM, Zhu HJ, Yuan HZ, et al. Guang Pu Xue Yu Guang Pu Fen Xi 22(1), 36-8, (2002)

Loading...


Tris(spiroborate)-Type Anionic Nanocycles Danjo, H., et al. Chem. Asian J. 7(7), 1529-32, (2012)

Loading...


Beil. 6,IV,6564

Aldrich MSDS 1, 710:C / Corp MSDS 1 (1), 1278:C / FT-IR 1 (1), 1112:B / FT-IR 2 (2), 1899:A / FT-NMR 1 (2), 312:B / IR-Spectra (3), 669:A / IR-Spectra (2), 597:B / NMR-Reference 2 (1), 911:C / RegBook 1 (1), 1305:E / Sax 6, 1972 / Sigma FT-IR 1 (2), 477:A / Structure Index 1, 199:D:4

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?