Synonym:
3-
CAS Number 153719-23-4
Empirical Formula (Hill Notation) C8H10ClN5O3S
Molecular Weight 291.71
Beilstein Registry Number 8491021
MDL number MFCD03792862
PubChem Substance ID 24863231
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?