Synonym: Ammonium iron(II) sulfate solution
CAS Number 10045-89-3
MDL number MFCD00051039
| Related Categories | Analytical Reagents, Analytical/Chromatography, By Reference Material, Most common compounds for volumetric titration, Reference Material Potassium permanganate, |
| grade | for COD determination |
| concentration | 0.06 M (NH4)2Fe(SO4)2 |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?