Synonym: Barium chloride solution
CAS Number 10361-37-2
Empirical Formula (Hill Notation) BaCl2
Molecular Weight 208.23
MDL number MFCD00003445
PubChem Substance ID 24863733
Popular Documents: FTNMR (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, By Reference Material, Reference Material Silver nitrate, Salt Concentrates, |
| packaging | pkg of 0.05 mol (12.214 g BaCl2 · 2H2O) |
| concentration | 0.05 M BaCl2 (0.1N) |
Visit our Titration Center to learn more.
concentration after dilution to 1L: 0.05mol/L (0.1N); amount of substance 0.05mol (12.214 g BaCl2.2H2O)
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?