Synonym:
(Ethylenedinitrilo)
CAS Number 139-33-3
Linear Formula [-CH2N(CH2CO2Na)CH2CO2H]2
Molecular Weight 336.21
Beilstein Registry Number 3822669
EC Number 205-358-3
MDL number MFCD00070672
PubChem Substance ID 24863752
| Related Categories | Analytical Reagents, Analytical/Chromatography, By Reference Material, Complexometric Concentrates (IDRANAL), Essential Chemicals, |
| grade | for complexometry |
| quality | (37.224 g C10H14N2Na2O8•2H2O, 0.1 mol) |
| packaging | pkg of 0.1 mol |
| concentration | 0.1 M EDTA-Na2 |
Visit our Titration Center to learn more.
concentration after dilution to 1L: 0.1mol/L; amount of substance 0.1mol (37.224 g C10H14N2Na2O8 · 2H2O)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?