Synonym: Potassium permanganate solution
CAS Number 7722-64-7
Empirical Formula (Hill Notation) KMnO4
Molecular Weight 158.03
MDL number MFCD00011364
PubChem Substance ID 24863802
| Related Categories | Analytical Reagents, Analytical/Chromatography, Most common compounds for volumetric titration, Potassium permanganate solution, Salt Concentrates, |
| packaging |
pkg of |
| concentration | 1/50 M KMnO4 (0.1N) |
Visit our Titration Center to learn more.
concentration after dilution to 1L: 1/50mol/L (0.1N); amount of substance 1/50mol (3.161 g KMnO4)
glass ampoule
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?