• USA Home
  • 38294 - Sulfuric acid concentrate

EMAIL THIS PAGE TO A FRIEND
38294 Fluka

Sulfuric acid concentrate

for 1 L standard solution, 0.5 M H2SO4 (1.0 N)

Synonym: Sulfuric acid solution

  • CAS Number 7664-93-9

  • Linear Formula H2SO4

  • Molecular Weight 98.08

  •  Beilstein Registry Number 2037554

  •  MDL number MFCD00064589

  •  PubChem Substance ID 24863925

Purchase

Description

General description

Visit our Titration Center to learn more.

concentration after dilution to 1L: 0.5mol/L (1.0N); amount of substance 0.5mol (49.039 g H2SO4)

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2796 8 / PGII
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?