EMAIL THIS PAGE TO A FRIEND

38300 Fluka

Diisopropylamine

purum, ≥98.0% (GC)

Synonym: DIPA

  • CAS Number 108-18-9

  • Linear Formula (CH3)2CHNHCH(CH3)2

  • Molecular Weight 101.19

  •  Beilstein Registry Number 605284

  •  EC Number 203-558-5

  •  MDL number MFCD00008862

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

vapor density   3.5 (vs air)
vapor pressure   50 mmHg ( 20 °C)
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   599 °F
expl. lim.   8.5 %
refractive index   n20/D 1.392(lit.)
  n20/D 1.393
bp   84 °C(lit.)
mp   −61 °C(lit.)
density   0.722 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Diisopropylamine

≥99.5%

Diisopropylamine

puriss. p.a., ≥99.0% (GC)

Diisopropylamine

analytical standard

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Hazard Codes (Europe) 
F,C
Risk Statements (Europe) 
Safety Statements (Europe) 
16-26-36/37/39-45
RIDADR 
UN 1158 8(3) / PGII
WGK Germany 
2
RTECS 
IM4025000
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Merck 14,3196

Beil. 4,IV,510

FT-IR 1 (1), 296:C / FT-NMR 1 (1), 474:C / RegBook 1 (1), 315:G / Structure Index 1, 43:B:4

Related Products

replaced by

Product #

Description

Add to Cart

471224 Diisopropylamine, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?