38520 Fluka

2,3-Dimercapto-1-propanol

for complexometry, for spectrophotometric det. of Cu, ≥98.0%

DOWNLOAD MSDS (PDF)

Synonym: BAL, British anti-Lewisite, DMP, Dimercaprol, Dithioglycerol

  • CAS Number 59-52-9

  • Linear Formula HOCH2CH(SH)CH2SH

  • Molecular Weight 124.23

  •  Beilstein Registry Number 1732058

  •  EC Number 200-433-7

  •  MDL number MFCD00004864

  •  PubChem Substance ID 24864016

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Reagents, Analytical Standards, Analytical/Chromatography, Complexometric Titration Reagents (EDTA Titration),
grade   for complexometry
assay   ≥98.0% (GC)
  ≥98.0%
quality   for spectrophotometric det. of Cu
refractive index   n20/D 1.573(lit.)
  n20/D 1.574
bp   120 °C/15 mmHg(lit.)
density   1.239 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

General description

Visit our Titration Center to learn more.

Other Notes

Chelator for heavy metals; used as antidote to heavy metal poisoning (arsenic, gold, mercury, etc)1,2; Reagent for extracting Cu(II)3; Semi-quantitative det. of Cu(II)4

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?