• USA Home
  • 38610 - 2,3-Dimethoxybenzaldehyde

EMAIL THIS PAGE TO A FRIEND

38610 Fluka

2,3-Dimethoxybenzaldehyde

purum, ≥97.0% (HPLC)

Purchase

Properties

grade   purum
assay   ≥97.0% (HPLC)
bp   137 °C/12 mmHg(lit.)
mp   48-52 °C(lit.)
  49-53 °C

Description

Other Notes

This product has been replaced by D130206-ALDRICH | 2,3-Dimethoxybenzaldehyde 98%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 8,IV,1747

Aldrich MSDS 1, 728:B / Corp MSDS 1 (1), 1306:A / FT-IR 1 (2), 114:A / FT-IR 2 (2), 2534:D / FT-NMR 1 (2), 947:C / IR-Spectra (2), 802:B / IR-Spectra (3), 916:D / NMR-Reference 2 (2), 112:B / RegBook 1 (2), 1721:A / Sax 6, 2722 / Sigma FT-IR 1 (2), 484:C / Structure Index 1, 273:D:4 / Vapor Phase 3, 1294:C

Related Products

replaced by

Product #

Description

Add to Cart

D130206 2,3-Dimethoxybenzaldehyde, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?