Synonym:
1-
CAS Number 555-66-8
Empirical Formula (Hill Notation) C17H24O3
Molecular Weight 276.37
Beilstein Registry Number 2056098
MDL number MFCD01736094
| Related Categories | Aldehydes and Ketones, Alphabetical, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | analytical standard |
| assay | ≥90.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?