39760 Fluka

2,3-Dimethylbutane

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: Diisopropyl

  • CAS Number 79-29-8

  • Linear Formula (CH3)2CHCH(CH3)2

  • Molecular Weight 86.18

  •  Beilstein Registry Number 1730737

  •  EC Number 201-193-6

  •  MDL number MFCD00008925

  •  PubChem Substance ID 24864705

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, D,
grade   analytical standard
vapor density   3 (vs air)
vapor pressure   7.41 psi ( 37.7 °C)
assay   ≥99.5% (GC)
autoignition temp.   788 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   ~7.7 %
refractive index   n20/D 1.375(lit.)
  n20/D 1.375
bp   58 °C(lit.)
mp   −129 °C(lit.)
density   0.662 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

2,2-Dimethylbutane

analytical standard

2,2-Dimethylbutane

≥99.0% (GC)

2-Methylpentane

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?