Synonym:
(R)
CAS Number 31886-58-5
Empirical Formula (Hill Notation) C14H19FeN
Molecular Weight 257.15
MDL number MFCD00066273
PubChem Substance ID 24865081
Popular Documents: FTNMR (PDF)
| Related Categories | Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Other Chiral Ligands |
| grade | purum |
| assay | ≥98.0% (T) |
| optical activity | [α]20/D +14±1°, c = 1.5% in ethanol |
| color | dark brown |
| refractive index |
n |
| bp | 120-121 °C/0.7 mmHg(lit.) |
| density | 1.222 g/mL at 20 °C(lit.) |
This chiral ferrocene derivative can be stereoselectively lithiated and derivatized; synthesis of chiral ferrocenyl phosphines, ligands for rhodium(I) 1,2
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?