• USA Home
  • 40834 - Actinomyces Broth, Vegitone

EMAIL THIS PAGE TO A FRIEND

40834 Fluka

Actinomyces Broth, Vegitone

for microbiology

Synonym: Vegitone Actinomyces Broth

Purchase

Properties

Related Categories A - M, Actinomycetes, All Liquid Media/Broths, Clinical Media, Media,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   ammonium sulfate, 1 g/L
  calcium chloride, 0.02 g/L
  L-cysteine hydrochloride, 1 g/L
  dextrose, 5 g/L
  magnesium sulfate, 0.2 g/L
  monopotassium phosphate, 15 g/L
  sodium chloride, 5 g/L
  starch, soluble, 1 g/L
  tryptone (vegetable), 4 g/L
  tryptose (vegetable), 10 g/L
  vegetable infusion powder, 10 g/L
  yeast extract, 5 g/L
final pH   7.2±0.2 (25 °C)
suitability   Actinomycetes
  Streptomyces

Description

Application

Actinomyces media are recommended for the cultivation and maintenance of the anaerobic Actinomyces species. Media from the Vegitone-line do not contain any animal derived material.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

for microbiology

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?