41008 Fluka

Ethylenediamine

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 1,2-Diaminoethane

  • CAS Number 107-15-3

  • Linear Formula NH2CH2CH2NH2

  • Molecular Weight 60.10

  •  Beilstein Registry Number 605263

  •  EC Number 203-468-6

  •  MDL number MFCD00008204

  •  PubChem Substance ID 24865689

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Aliphatics, Alpha Sort, Alphabetical Index of Analytical Standards, Amines, Analytical Standards,
grade   analytical standard
  for GC
vapor density   2.07 (vs air)
vapor pressure   10 mmHg ( 20 °C)
assay   ≥99.8% (GC)
autoignition temp.   716 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   16 %
refractive index   n20/D 1.456
  n20/D 1.4565(lit.)
bp   118 °C(lit.)
mp   8.5 °C(lit.)
density   0.899 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Ethylenediamine

BioXtra

Ethylenediamine

puriss. p.a., absolute, ≥99.5% (GC)

Ethylenediamine

purified by redistillation, ≥99.5%

Ethylenediamine solution

technical, 75-80%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?