EMAIL THIS PAGE TO A FRIEND

41132 Fluka

Fluopicolide

PESTANAL®, analytical standard

Synonym: 2,6-Dichloro-N-[3-chloro-5-(trifluoromethyl)-2-pyridylmethyl]benzamide

  • CAS Number 239110-15-7

  • Empirical Formula (Hill Notation) C14H8Cl3F3N2O

  • Molecular Weight 383.58

  •  MDL number MFCD09751509

Purchase

Properties

Related Categories All Fungicides, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, E-G,
grade   analytical standard
product line   PESTANAL®
form   neat
format   neat

Description

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability


Explore the New Analytical Standards Portal

Customers Also Viewed

Cyazofamid

PESTANAL®, analytical standard

Fenamidone

PESTANAL®, analytical standard

Mandipropamid

PESTANAL®, analytical standard

Propamocarb

PESTANAL®, analytical standard

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?