Synonym: DERA Aldolase from Lactobacillus plantarum
CAS Number 9026-97-5
MDL number MFCD03095921
| form | powder |
| color | slightly yellow |
| storage temp. | −20°C |
Enzyme for the stereoselective synthesis of carbohydrates by multi-substrate aldol reactions1,2
1 U corresponds to the amount of enzyme which converts 1 μmol 2-deoxy-
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?