41270 Fluka

Violet Red Bile Lactose Dextrose Agar

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: VRB Lactose Dextrose Agar, Violet Red Bile Lactose Glucose Agar

Properties

Related Categories All Food Control Media (alphabetical), All Solid Media/Agars, Citrobacter, Enterobacteriaceae, Enterococcus,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   agar, 15.0 g/L
  bile salts, 1.5 g/L
  crystal violet, 0.002 g/L
  dextrose monohydrate, 10 g/L
  lactose monohydrate, 10.0 g/L
  neutral red, 0.03 g/L
  pancreatic digest of gelatin, 7.0 g/L
  sodium chloride, 5.0 g/L
  yeast extract, 3.0 g/L
pH   7.4±0.2 (25 °C)
suitability   selective and differential for Citrobacter
  selective and differential for Enterobacteriaceae
  selective and differential for Enterococcus
  selective and differential for Escherichia coli and Coliforms
  selective and differential for Klebsiella
  selective and differential for Proteus
  selective and differential for Salmonella
  selective and differential for Shigella

Description

General description

A selective Agar for the detection of Enterobacteriaceae according the European Pharmacopeia. he medium is specially used in the recovery of process-stressed bacteria using a progressive enrichment technique.

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?