414689 Fluka

Acetone

capillary GC grade, ≥99.9%

DOWNLOAD MSDS (PDF)
  • CAS Number 67-64-1

  • Linear Formula CH3COCH3

  • Molecular Weight 58.08

  •  Beilstein Registry Number 635680

  •  EC Number 200-662-2

  •  MDL number MFCD00008765

  •  PubChem Substance ID 24865952

Properties

Related Categories Amber Glass Bottles, GC Capillary, GC Solvents, Solvent Bottles, Solvent Packaging Options,
grade   capillary GC grade
vapor density   2 (vs air)
vapor pressure   184 mmHg ( 20 °C)
assay   ≥99.9%
expl. lim.   13.2 %
impurities   ≤1 ng/mL organic residue (GC/FID, as hexadecane)
evapn. residue   <0.0001%
halogenated residue (GC/ECD)   <2 ng/L (as lindane)
refractive index   n20/D 1.359(lit.)
bp   56 °C/760 mmHg(lit.)
mp   −94 °C(lit.)
density   0.791 g/mL at 25 °C(lit.)

Description

Application

Suitable for environmental analysis

Acetone′s luminesence intensity is dependent upon the solution components 1. The absorption of UV light by acetone, results in its photolysis and the production of radials 1.

Packaging

4×4 L in glass bottle

Price and Availability

Customers Also Viewed

Acetone
650501Acetone

CHROMASOLV® Plus, for HPLC, ≥99.9%

Acetone
270725Acetone

CHROMASOLV®, for HPLC, ≥99.9%

Acetone
34850Acetone

CHROMASOLV®, for HPLC, ≥99.8%

Acetone
439126Acetone

CHROMASOLV®, for HPLC, ≥99.9%

Acetone
179124Acetone

ACS reagent, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?