414816 Fluka

Methanol

for GC/MS analysis of volatile organics, ≥99.9%

DOWNLOAD MSDS (PDF)

Synonym: Methyl alcohol

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chromatography Reagents & Solvents, GC Solvents, Methanol,
vapor density   1.11 (vs air)
vapor pressure   410 mmHg ( 50 °C)
  97.68 mmHg ( 20 °C)
assay   ≥99.9% (GC)
  ≥99.9%
autoignition temp.   725 °F
quality   for GC/MS analysis of volatile organics
expl. lim.   36 %
impurities   ≤0.2% water
evapn. residue   <0.5 mg/L
refractive index   n20/D 1.329(lit.)
bp   64.7 °C(lit.)
  64.7 °C
mp   −98 °C(lit.)
density   0.791 g/mL at 25 °C(lit.)
  0.791 g/mL at 25 °C

Description

Application

Suitable for volatile organics analysis by GC-MS

Packaging

1 L in glass bottle

Price and Availability

Customers Also Viewed

Methanol

CHROMASOLV®, for HPLC, ≥99.9%

Methanol

LC-MS Ultra CHROMASOLV®, tested for UHPLC-MS

Methanol

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)

Methanol

LC-MS CHROMASOLV®

Methanol
414719Methanol

capillary GC grade, ≥99.9%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?