• USA Home
  • 42376 - Violet Red Bile Lactose Agar, Vegitone

EMAIL THIS PAGE TO A FRIEND

42376 Fluka

Violet Red Bile Lactose Agar, Vegitone

for microbiology

Synonym: Crystal violet-Neutral red-Bile Agar, Vegiton, VRB-Agar, Vegitone, Vegitone Violet Red Bile Agar

Purchase

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Control Media for Fish, Enterobacter,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   agar, 12.0 g/L
  crystal violet, 0.002 g/L
  lactose, 10.0 g/L
  neutral red, 0.03 g/L
  peptone (vegetable), 7.0 g/L
  sodium chloride, 5.0 g/L
  synthetic detergent, 1.5 g/L
  yeast extract, 3.0 g/L
final pH   7.4±0.2 (25 °C)
suitability   selective and differential for Enterobacter
  selective and differential for Escherichia coli and Coliforms
  selective and differential for Salmonella

Description

Application

Violet Red Bile Agar Vegitone is a selective media used for the detection and enumeration of coliform organisms from water and food. This medium contains no animal derived peptone and bile salt.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

for microbiology

for microbiology

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Differentiation and Identification Media for Salmonella

Salmonella contamination is the second leading cause of food-borne illness worldwide. Controlling outbreaks of Salmonella is an important task for food regulators, restaurants and the food industry i...
Jvo Siegrist
Microbiology Focus Edition 1.3
Keywords: Biochemistry, Fermentation, Food & Beverage, Indicators, Microbiology

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?