42414 Fluka

2′,3′-Dideoxyguanosine 5′-triphosphate sodium salt solution

puriss. p.a., for MALDI MS, ≥98.0% (HPLC), 10.0-11.0 (UV)

DOWNLOAD MSDS (PDF)

Synonym: ddGTP

  • Empirical Formula (Hill Notation) C10H15N5NaO12P3

  • Molecular Weight 513.16

  •  MDL number MFCD00079152

  •  PubChem Substance ID 24866554

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Bioactive Small Molecules, Biochemicals and Reagents, Cell Biology,
grade   for MALDI MS
  puriss. p.a.
assay   ≥98.0% (HPLC)
packaging   pkg of 1 μmol
concentration   10.0-11.0 (UV)
impurities   ≤1.5% ddNDP (HPLC)
suitability   corresponds for UV test
storage temp.   −20°C
Gene Information   human ... HRAS(3265)

Price and Availability

Customers Also Viewed

2′,3′-Dideoxyadenosine 5′-triphosphate sodium salt solution

puriss. p.a., for MALDI MS, ≥98.0% (HPLC)

3′-Deoxythymidine 5′-triphosphate sodium salt solution

BioChemika, puriss. p.a., for MALDI MS, ≥98.0% (HPLC)

for MALDI MS

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?