42429 Fluka

p-Cresol

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 4-Methylphenol

  • CAS Number 106-44-5

  • Linear Formula CH3C6H4OH

  • Molecular Weight 108.14

  •  Beilstein Registry Number 1305151

  •  EC Number 203-398-6

  •  MDL number MFCD00002376

  •  PubChem Substance ID 24866562

  • Popular Documents:  FTIR (PDF)  

Properties

Related Categories A-F, Alcoholic & Nonalcoholic Beverages, Alcohols, All Alphabetically Sorted, Alpha Sort,
grade   analytical standard
vapor density   3.72 (vs air)
vapor pressure   1 mmHg ( 20 °C)
assay   ≥99.7% (GC)
autoignition temp.   1038 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   1 %
  1.1 %, 150 °F
bp   202 °C(lit.)
mp   32-34 °C(lit.)
  32-35 °C
density   1.034 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

4-Methylphenol

analytical standard

<I>m</I>-Cresol

analytical standard

<I>p</I>-Cresol

puriss. p.a., ≥99.0% (GC)

<I>p</I>-Cresol

purum, ≥98.0% (GC)

<I>p</I>-Cresol
W233706p-Cresol

≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?