Synonym:
(±)-1,2-Dipalmitin,
1,2-Dihexadecanoyl-rac-glycerol,
rac-Glycerol 1,2-dipalmitate,
CAS Number 40290-32-2
Empirical Formula (Hill Notation) C35H68O5
Molecular Weight 568.91
Beilstein Registry Number 1809777
EC Number 254-870-3
MDL number MFCD00042874
PubChem Substance ID 24866640
| Related Categories | Analytical Reagents, Analytical/Chromatography, Biochemicals and Reagents, Building Blocks, C12 to C63, |
| assay | ≥99.0% (after transesterification, GC) |
| mp | 64-66 °C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?